Learn More
FSI North AmericaSupplier Diversity Partner DAT2525S Portable Pneumatic Hazmat Decon Shower DFS Item
Manufacturer:  FSI North AmericaSupplier Diversity Partner DAT2525S
approx. 7' W X 13' L X 8' H, with 4 ft. Undress/5 ft. Shower/4 ft. Redress areas. A First Responder Ambulatory Decon shower system with 6 spray nozzles and 2 trigger gun with coiled hoses, 90 sq. ft., 180 lbs. Inflator/deflator included.
Catalog No. 50-195-2447