Learn More
Dawson Calibration Gas Cylinders
Calibration mixes in convenient cylinders
Manufacturer:  Dawson 6000050000
 View more versions of this product
Catalog No. 19-809-147
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Oxygen 20.9%, carbon monoxide 50ppm, hydrogen sulfide 25ppm, LEL 50%, nitrogen
Multigas:Oxygen, Carbon Monoxide, Hydrogen Sulfide, LEL, Nitrogen
58L
15.32 gal.
Calibration Gas Cylinder