Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAX-1 / NR0B1 Mouse anti-Human, Mouse, Rat, Clone: 1DA-2F4, Active Motif™

Mouse Monoclonal Antibody
Supplier: Active Motif 39983

Description
This gene encodes a protein that contains a DNA-binding domain. The encoded protein acts as a dominant-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in this gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism.Specifications
DAX-1 / NR0B1 | |
Monoclonal | |
1 mg/mL | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
P51843, P70503, Q61066 | |
NR0B1 | |
This DAX-1 / NR0B1 antibody was raised against a peptide corresponding to amino acids 135-166 of human DAX-1 / NR0B1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
1DA-2F4 | |
Unconjugated | |
NR0B1 | |
adrenal hypoplasia, congenital homolog; AHC; Ahch; AHX; BOS_26183; DAX; DAX 1; DAX1; DAX-1; DSS; DSS-AHC critical region on the X chromosome protein 1; GTD; HHG; NR0B1; NROB1; nuclear hormone receptor; Nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1; orphan nuclear receptor DAX-1; R0B1; SRXY2 | |
Mouse | |
Protein A | |
RUO | |
11614, 190, 58850 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction