Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daxx Mouse anti-Human, Clone: PCRP-DAXX-8B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30783820UG

 View more versions of this product

Catalog No. NB123150


Add to cart

Description

Description

Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

Daxx
PCRP-DAXX-8B7
Unconjugated
10 mM PBS with 0.05% BSA
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246
Recombinant full-length human Daxx protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Peptide Array
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Purified
Mouse
IgG2b Kappa
20 μg
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary
1616
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit