Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Daxx Mouse anti-Human, Clone: PCRP-DAXX-8B7, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Daxx |
---|---|
Clone | PCRP-DAXX-8B7 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB123150
|
Novus Biologicals
NBP30783820UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Daxx Monoclonal specifically detects Daxx in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
Daxx | |
0.2 mg/ml | |
Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
10 mM PBS with 0.05% BSA | |
1616 | |
IgG2b κ | |
Protein A or G purified |
PCRP-DAXX-8B7 | |
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
Recombinant full-length human Daxx protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title