Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Daxx Mouse anti-Human, DyLight 405, Clone: PCRP-DAXX-5G11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314025V
This item is not returnable.
View return policy
Description
Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Daxx | |
Monoclonal | |
DyLight 405 | |
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246 | |
Recombinant full-length human Daxx protein (Uniprot: Q9UER7) | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
1616 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PCRP-DAXX-5G11 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title