Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAZAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB30695

Catalog No. 89-033-035


Add to cart

Description

Description

Rabbit polyclonal antibody raised against partial recombinant human DAZAP1.

  • High specificity
  • High reactivity
Specifications

Specifications

DAZAP1
Unconjugated
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20-1:50) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Q96EP5
MGC19907
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Mouse, Rat
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against partial recombinant human DAZAP1.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
Liquid
DAZAP1
Recombinant protein corresponding to human DAZAP1.
Antigen affinity purification
RUO
Primary
26528
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)