Learn More
Medchemexpress LLC DB2115 tertahydrochloride | 1366126-19-3 | 99.1% | 704.48 g/mol | C32H34Cl4N8O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY124676A10MG
Encompass_Preferred
DB2115 tertahydrochloride is a tetrahydrochloride salt small-molecule inhibitor of the transcription factor PU.1, described as highly selective with reported nanomolar potency. It is supplied as a high-purity solid intended for in vitro biochemical and cell-based research into PU.1-mediated transcription, tumor cell proliferation, and apoptosis.
  • Highly selective PU.1 inhibition with reported nanomolar potency.
  • High chemical purity suitable for sensitive assays.
  • Solid form enables accurate weighing and dosing.
  • Available in small-scale research quantities for laboratory use.
  • Recommended storage under sealed, dry conditions to preserve stability.
Catalog No. 50-002-37167