Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-SALA™ Delta™ Construction Style Positioning Harness
Featuring coated, corrosion-resistant hardware, water-repellant and abrasion-resistant webbing and a build quality that is second to none. Also incorporates a web spreading design, positioning webbing wider apart on shoulder straps to eliminate abrasion on your neck.
Supplier: 3M™ 7100232003
Description
Specifications
190 kg | |
Repel™ Polyester | |
Tongue buckle | |
420 lb. |
Large | |
Back, Side | |
5.6 lb. | |
n/a |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction