Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-SALA™ ExoFit NEX™ Vest-Style Retrieval Harness
3M™ DBI-SALA™ ExoFit NEX™ Vest-Style Retrieval Harness features an aluminum back, shoulder D-rings, locking quick connect buckle leg straps, and comfort padding
Supplier: 3M™ 7100188757
Description
- Hybrid shoulder, back and leg comfort padding
- Stand-up lightweight aluminum back D-ring
- Lightweight aluminum shoulder D-rings
- Web-locking quick-connect chest and leg straps
- Repel™ webbing
- Loops for body belt
- Protective shoulder caps
- Built-in lanyard keepers
- Revolving vertical torso buckle
- Impact Indicator
- Integrated trauma straps
- Built-in reflective materials
- Protected labels, equipped with 3M™ Connected Safety ID (CSID)
Specifications
Duo-Lok™ Quick Connect | |
420 lb. | |
OSHA 1910.66, ANSI Z359.1, ANSI Z359.4, ANSI A10.32, OSHA 1926.502 | |
Medium |
Back, Shoulder | |
Vest-Style Retrieval Harness | |
Repel™ Polyester | |
4.3 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction