Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ DBI-SALA™ ExoFit™ Vest-Style Retrieval Harness
Back and shoulder D-rings, quick connect buckle leg straps, built-in comfort padding
Supplier: 3M™ 1108753
Description
- Built-in shoulder, back and leg comfort padding
- Back D-ring
- Shoulder D-rings
- Quick connect chest and leg straps
- Soft, lightweight webbing
- Loops for body belt
- Anti-tear ballistic nylon
- Built-in reflective piping
- Parachute torso adjusters
- Protected labels, equipped with i-Safe™
Specifications
Back, Shoulder | |
420 lb. | |
Polyester | |
Vest-Style Retrieval Harness |
190 kg | |
Quick Connect | |
6000 lb. | |
Large |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction