Learn More
DBI/Sala™ Mounted Recovery System: Winches
Manufacturer:  DBI Sala Inc. 8101002
 View more versions of this product
Catalog No. 18-999-1150
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Salalift Winch
Silver
Mounted recovery system
60 ft.
0.25 in. stainless steel cable
60 ft. stainless steel cable, mounting bracket, triple braking system, quick mount bracket, carrying bag
OSHA 1910.28, 1926.451, OSHA 1910.146, ANSI Z117.1
62 lb.