Learn More
3M™ DBI-SALA™ Nano-Lok™ Twin-Leg Quick Connect Self Retracting Lifeline - Web
Designed for ease-of-use and is ideal for direct connection to most harnesses. The extremely compact and lightweight design is virtually unnoticeable on your back, staying out of the way and making it ideal as a lanyard replacement.
Manufacturer:  3M™ 3101288
 View more versions of this product
Catalog No. 19-070-705
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Twin-leg 100% tie-off design
  • Built-in connector for twin-leg SRL direct attachment to harness
  • 6 ft. (1.8 m) twin-leg lifelines with 3/4 in. (19 mm) Dyneema® fiber and polyester web
  • Self-locking rebar hook on each lifeline
  • Extremely compact, lightweight design
  • Tough and impact resistant thermoplastic housing
  • Swiveling anchorage loops
  • Quick activating speed sensing brake systems
  • High capacity external energy absorbers and impact indicators
  • i-Safe™ equipped
  • Specifications
    Specifications
    Construction, General Industrial, Mining, Oil and Gas, Transportation, Utilities, Wind Energy
    Built-in connector for twin-leg SRL direct attachment to harness (fixed D-ring)
    ANSI Z359.14, ANSI Z359.1
    190 kg
    1.8 m
    Safety
    Web, Zinc Plated Steel, Nylon Thermoplastic
    420 lbs.
    6 ft.
    6.3 lbs.