Learn More
3M™ DBI-SALA™ R550 Rescue System
DBI-SALA™ R550 Rescue System designed specifically for rescues from drilling and service rigs.
Manufacturer:  3M™ 70804512252
Catalog No. 19-072-169
Add to cart
Description
Description
  • Rescue System is intended for the oil and gas drilling and service industry
  • Pictorial label cards provided for EA pocket allow for quick location identification and proper product use
  • Rescue System contents are organized and secured within designated pockets. Fall protection for tools incorporated for all content within the bag
  • Bag is compact and incorporates versatile pocket storage
  • Bag has a rated lifting point integrated on top
  • Rescue System is easy to use
  • Bag incorporates padded shoulder straps to provide comfort while carrying
  • The ridged bag design keeps bag aligned with the worker while climbing structure
Specifications
Specifications
Oil and Gas Rescue System
Two 6 ft. Web Slings, Two 1.75 ft. Web Slings, Four 10 lb. Tool Tethers, One 2 in. Gate Opening Carabiners, Four Steel Carabiners, One Micro Pulley, One RA Edge Protector, One Bungee Tether, One Cable Grab, One Web Grab, One 200 in.R550 Rescue Unit
620 lb.
200 ft.
29.5 lb.
Kernmantle Rope
R550: ANSI Z359.4; R550: EN 341:1993; R550: CSA Z259.2.3
281 kg
61 m
13.38 kg