Learn More
DBI/Sala™ Sealed Self-Retracting Lifeline
Removes the danger of trailing lines
Manufacturer:  DBI Sala Inc. 3403500
 View more versions of this product
Catalog No. 18-723
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Self retracting lifeline
Spring tensioned 5mm Galvanized Stainless Steel cable
310 lb.
85 ft.
44 lb.
Galvanized Steel
ANSI Z359.1-1992, ANSI A10.14-1991, OSHA
140.61kg
25.9m
19.95kg