Learn More
3M™ DBI-SALA™ Reusable Standing Seam Roof Anchor
Standing seam roof anchor with swiveling SRL bracket, telescoping adjustment fits 24, 30, 32 and 36 in. seam spacing (61, 76.2, 81.3, 91.4 cm).
Manufacturer:  3M™ 2103675
Catalog No. 19-809-350
Add to cart
Description
Description
  • Installs to flat or sloped standing seam roofs
  • Built-in non-penetrating clamp design
  • Telescoping adjustment legs fit 24, 30, 32 and 36 in. seam spacing (61, 76.2, 81.3, 91.4 cm)
  • Built-in swiveling self retracting lifeline bracket (order SRL separately)
  • Durable and corrosion resistant construction
  • Compact and lightweight design
Specifications
Specifications
26.4 x 15 in.
Black
OSHA 1910.140, OSHA 1926.502
Reusable Standing Seam Roof Anchor
24 in., 30 in., 32 in. and 36 in. seam spacing
48 lbs.