Learn More
3M™ DBI-SALA™ Reusable Swiveling Quad-Pod Roof Anchor with SRL
Reusable swiveling quad-pod roof anchor with 30 ft. (9.1m) Ultra-Lok™ self retracting lifeline and fasteners for flat or sloped wood roofs.
Manufacturer:  3M™ 3505694
Catalog No. 19-809-393
Add to cart
Description
Description
  • Installs to flat or sloped wood roofs
  • Fast and easy nail or screw installation - Fastener kit included
  • Unique tall design elevates lifeline from roof surface
  • Built-in swiveling self retracting lifeline bracket
  • Includes 30 ft. (9.1m) Ultra-Lok™ self retracting lifeline
  • Durable and corrosion resistant construction
Specifications
Specifications
10 x 0.20 in.
Reusable Swiveling Quad-Pod Roof Anchor with SRL
Ultra-Lok™ Self retracting lifeline
30 ft.
14 lbs.
5000 lbs.
Black,Yellow Zinc
OSHA 1910.140, OSHA 1926.502
914.4 cm