Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ DC-LAMP Human Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$344.00 - $1298.00
Specifications
Concentration | 0.5mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 27074 |
Molecular Weight | TMW: 38.8kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25229905
|
Novus Biologicals™
NBP2522990.05MG |
0.05mg |
Each for $344.00
|
|
NBP25229925
|
Novus Biologicals™
NBP2522990.25MG |
0.25mg |
Each for $1,298.00
|
|
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 28-381 of Human DC-LAMP The Recombinant Human DC-LAMP Protein is derived from Baculovirus. The Recombinant Human DC-LAMP Protein has been validated for the following applications: SDS-Page.Specifications
0.5mg/mL | |
Phosphate buffered saline (pH 7.4) containing 10% glycerol | |
TMW: 38.8kDa | |
Baculovirus | |
CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
Unlabeled |
SDS-Page | |
27074 | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
DC-LAMP | |
Yes |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title