Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-LAMP Rabbit anti-Human, Clone: 7, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | DC-LAMP |
---|---|
Clone | 7 |
Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100 |
Applications | Immunofluorescence |
Classification | Monoclonal |
Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DC-LAMP | |
Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 | |
Recombinant Human DC-LAMP protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
7 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Dendritic Cell Markers | |
0.2 um filtered solution in PBS | |
27074 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title