Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SCRIPT/ZNF366 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | DC-SCRIPT/ZNF366 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180414
|
Novus Biologicals
NBP180414 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
DC-SCRIPT/ZNF366 Polyclonal specifically detects DC-SCRIPT/ZNF366 in Human samples. It is validated for Western Blot.Specifications
DC-SCRIPT/ZNF366 | |
Polyclonal | |
Rabbit | |
NP_689838 | |
167465 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
FLJ39796, zinc finger protein 366 | |
ZNF366 | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title