Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN/CD209 Mouse anti-Human, Alexa Fluor™ 594, Clone: UW60.1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221791AF594

Catalog No. NP221791A94


Add to cart

Description

Description

DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

DC-SIGN/CD209
UW60.1
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
CD209
CD209-His-tagged fusion protein.
Protein A purified
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary
30835.0
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody UW60.1 recognizes human CD209 (DC-SIGN), a 44 kDa transmembrane receptor expressed on the surface of dendritic cells and macrophages.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit