Learn More
DC-SIGN/CD209 Mouse anti-Human, Alexa Fluor™ 750, Clone: UW60.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221791AF750
Catalog No. NP221791A70
Add to cart
Description
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
DC-SIGN/CD209
UW60.1
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody UW60.1 recognizes human CD209 (DC-SIGN), a 44 kDa transmembrane receptor expressed on the surface of dendritic cells and macrophages.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD209
CD209-His-tagged fusion protein.
0.1 mL
Primary
30835
Human