Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Mouse anti-Human, Biotin, Clone: UW60.1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221791B
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DC-SIGN/CD209 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
CD209 | |
CD209-His-tagged fusion protein. | |
0.1 mL | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
30835 | |
Human | |
Purified |
Immunocytochemistry, Immunofluorescence | |
UW60.1 | |
Immunocytochemistry/Immunofluorescence | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody UW60.1 recognizes human CD209 (DC-SIGN), a 44 kDa transmembrane receptor expressed on the surface of dendritic cells and macrophages. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title