Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Mouse anti-Human, PE/Cy5.5, Clone: MM0239-4G10, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP212150PECY55
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry.Specifications
DC-SIGN/CD209 | |
MM0239-4G10 | |
Flow Cytometry | |
Conjugated, Purified | |
CD209 | |
Human DC-SIGN | |
Protein G purified | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
Primary | |
30835.0 |
Flow Cytometry | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title