Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Antibody (059), Alexa Fluor™ 350
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289331AF350
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
DC-SIGN/CD209 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
59 | |
Sandwich ELISA Detection | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DC-SIGN/CD209. | |
0.1 mL | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
30835 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction