Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGN/CD209 Rabbit anti-Human, Clone: 61, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | DC-SIGN/CD209 |
---|---|
Clone | 61 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot, ELISA.Specifications
DC-SIGN/CD209 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
Recombinant Human DC-SIGN/CD209 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
61 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Immunology, Virology Bacteria and Parasites | |
0.2 um filtered solution in PBS | |
30835 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title