Learn More
DC-SIGN/CD209 Rat anti-Mouse, Alexa Fluor 700, Clone: 902404, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB8345N100UG
Catalog No. FAB8345N100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
DC-SIGN/CD209
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q91ZX1
Cd209a
NS0 mouse myeloma cell line transfected with mouse DC-SIGN/CD209 Met1-Lys238 Accession # Q91ZX1
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse DC-SIGN/CD209 in direct ELISAs.
Flow Cytometry
902404
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Rat
IgG2b
RUO
Primary
30835
Mouse
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)