Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGN/CD209 Rat anti-Mouse, FITC, Clone: LWC06, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143328F

Catalog No. NBP143328F


Add to cart

Description

Description

DC-SIGN/CD209 Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry, Western Blotting.
Specifications

Specifications

DC-SIGN/CD209
LWC06
Flow Cytometry
Purified
CD209
The immunogen for this antibody was CIRE.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry
FITC
PBS with 0.05% Sodium Azide
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Rat
IgG2a
0.1 ml
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary
30835
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit