Learn More
DC-SIGN/CD209 Rat anti-Mouse, Janelia Fluor 549, Clone: LWC06, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143328JF549
Catalog No. NBP43328549
Add to cart
Description
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
DC-SIGN/CD209
LWC06
Flow Cytometry
Purified
CD209
The immunogen for this antibody was CIRE.
Protein A or G purified
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary
30835
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Rat
IgG2a
0.1mL
Store at 4°C in the dark.
Monoclonal
Mouse