Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGNR/CD299/CLEC4M Rabbit anti-Human, Alexa Fluor™ 750, Clone: 160000000, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252649AF750

Catalog No. NP22649A70


Add to cart

Description

Description

DC-SIGNR/CD299/CLEC4M Monoclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence.
Specifications

Specifications

DC-SIGNR/CD299/CLEC4M
160000000
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Conjugated, Purified
CLEC4M
CD299
Protein A or G purified
Adaptive Immunity, Immunology, Signal Transduction, Virology Bacteria and Parasites
Primary
10332.0
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human CD299, which is an oligomeric type II transmembrane protein with a C-type lectin extracellular domain, the expression of which is restricted to immature DC, macrophages in the lung, and endothelial cells in the liver. It binds ICAM-3 and ICAM-7 to mediate the interaction of DC with T lymphocytes and endothelial cells in the initial stages of immune response and in the migratory behavior of DC. CD299 also binds the gp120 protein of HIV and the E2 envelope protein of HCV, thereby playing a role in viral infection.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit