Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-SIGNR/CD299/CLEC4M Rabbit anti-Human, Clone: 160000000, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP288989

Catalog No. NB036580

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

DC-SIGNR/CD299/CLEC4M Monoclonal antibody specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications

Specifications

DC-SIGNR/CD299/CLEC4M
160000000
PBS
CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR
DC-SIGNR/CD299/CLEC4M
Protein A purified
RUO
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry
Unconjugated
Purified
Rabbit
IgG Kappa
100 μg
Adaptive Immunity, Immunology, Signal Transduction, Virology Bacteria and Parasites
Primary
10332
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit