Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DC-SIGNR/CD299/CLEC4M Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | DC-SIGNR/CD299/CLEC4M |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159181
|
Novus Biologicals
NBP159181 |
100 μL |
Each of 1 for $436.00
|
|
Description
DC-SIGNR/CD299/CLEC4M Polyclonal specifically detects DC-SIGNR/CD299/CLEC4M in Human samples. It is validated for Western Blot.Specifications
DC-SIGNR/CD299/CLEC4M | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
CD209L1, CD209Ldendritic cell-specific ICAM-3-grabbing nonintegrin 2, CD299, CD299 antigenMGC129964, C-type lectin domain family 4, member M, DC-SIGN2CD209 antigen-like protein 1, DC-SIGNRDC-SIGN-related protein, DCSIGNRMGC47866, Dendritic cell-specific ICAM-3-grabbing non-integrin 2, HP10347, liver/lymph node-specific ICAM-3 grabbing non-integrin, Liver/lymph node-specific ICAM-3-grabbing non-integrin, L-SIGN, LSIGNC-type lectin domain family 4 member M, mannose binding C-type lectin DC-SIGNR | |
CLEC4M | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q9H2X3 | |
10332 | |
Synthetic peptides corresponding to CLEC4M(C-type lectin domain family 4, member M) The peptide sequence was selected from the n terminal of CLEC4M. Peptide sequence MSDSKEPRVQQLGLLEEDPTTSGIRLFPRDFQFQQIHGHKSSTGCLGHGA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title