Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCAKD Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548901

Catalog No. PA548901

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Specifications

Specifications

DCAKD
Polyclonal
PBS with 0.09% sodium azide
Q8WVC6
Dephospho-CoA kinase domain-containing protein, DCAKD
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79877
Western Blot
Unconjugated
DCAKD
Liquid
DCAKD
KLH conjugated synthetic peptide between 51-80 amino acids from the N-terminal region of human DCAKD.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit