Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCB, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
6971-97-7
C14H10Cl2N2
277.148
XMOVWXSCYLINBJ-BUOZRGFLSA-N
3,3'-dichlorobenzylideneazine,benzaldehyde, 3-chlorophenyl methylene hydrazone
6861558
(E)-1-(3-chlorophenyl)-N-[(Z)-(3-chlorophenyl)methylideneamino]methanimine
C1=CC(=CC(=C1)Cl)C=NN=CC2=CC(=CC=C2)Cl
This item is not returnable. View return policy
6971-97-7
C14H10Cl2N2
277.148
XMOVWXSCYLINBJ-BUOZRGFLSA-N
3,3'-dichlorobenzylideneazine,benzaldehyde, 3-chlorophenyl methylene hydrazone
6861558
(E)-1-(3-chlorophenyl)-N-[(Z)-(3-chlorophenyl)methylideneamino]methanimine
C1=CC(=CC(=C1)Cl)C=NN=CC2=CC(=CC=C2)Cl

Allosteric modulator at mGlu5

Allosteric ligand for the metabotropic glutamate receptor mGlu5; displays neutral modulation. Does not affect agonist-stimulated mGlu5 responses, but blocks regulation of the receptor by other allosteric modulators such as DFB (Cat. No. 1625) and DMeOB (Cat. No. 1953).
Quantity 10mg
Formula Weight 277.15
Percent Purity >98%
Chemical Name or Material DCB

Recommended Storage : Desiccate at Room Temperature