Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCBLD2/ESDN Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093567AF488
Description
DCBLD2/ESDN Polyclonal antibody specifically detects DCBLD2/ESDN in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
DCBLD2/ESDN | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
CLCP11700055P21Rik, CUB, LCCL and coagulation factor V/VIII-homology domains protein 1, discoidin, CUB and LCCL domain containing 2, discoidin, CUB and LCCL domain-containing protein 2, Endothelial and smooth muscle cell-derived neuropilin-like protein, ESDNcoagulation factor V/VIII-homology domains protein 1 | |
Synthetic peptide corresponding to residues 402-419 [QDKIFQGNKDYHKDVRNN] of the human ESDN protein. | |
0.1 mL | |
Signal Transduction | |
131566 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction