Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCDC2 Polyclonal Antibody
DCDC2 Polyclonal Antibody
Supplier: Thermo Scientific PA556786
Description
DCDC2 Polyclonal Antibody for ICC/IF, IHC

Specifications
DCDC2 | |
Polyclonal | |
Unconjugated | |
DCDC2 | |
AW492955; Dcdc2; Dcdc2a; DFNB66; doublecortin domain containing 2; doublecortin domain containing 2a; doublecortin domain-containing protein 2; HGNC:18141; Kiaa1154; MGC54817; NPHP19; Protein RU2S; RU2; RU2S | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51473 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UHG0 | |
DCDC2 | |
Recombinant protein corresponding to Human DCDC2. Recombinant protein control fragment (Product #RP-94879). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction