Learn More
DCIR/CLEC4A Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0237-5P20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212257AF350
Catalog No. NP21227A30
Add to cart
Description
Description
DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
DCIR/CLEC4A
MM0237-5P20
Western Blot, Flow Cytometry
CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CLEC4A
Human CLEC4A
0.1 mL
Primary
50856