Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCIR/CLEC4A Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0237-5P20, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212257AF750
Description
DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
DCIR/CLEC4A | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CLEC4A | |
Human recombinant DCIR/CLEC4A (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry | |
MM0237-5P20 | |
Western Blot, Flow Cytometry | |
CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor | |
Mouse | |
Protein G purified | |
Immunology, Innate Immunity | |
50856.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title