Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCK Mouse anti-Human, Clone: GT7710, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527741

Catalog No. PIMA527741


Add to cart

Description

Description

Positive Control: HeLa, HepG2, 293T, A549, H1299, HCT116 Predicted Reactivity: Rhesus Monkey (96%), Bovine (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

DCK (Deoxycytidine kinase) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
Specifications

Specifications

DCK
Monoclonal
1 mg/ml
PBS with no preservative
P27707
deoxycytidine kinase
Mouse
IgG1
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1633
Western Blot
GT7710
Unconjugated
DCK
Liquid
DCK
Recombit protein encompassing a sequence within the center region of human DCK. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit