Learn More
DCK Mouse anti-Human, Clone: OTI16E12, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA502650
Catalog No. 50-167-3688
Add to cart
Description
Description
Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
Specifications
Specifications
DCK
Monoclonal
Unconjugated
DCK
Liquid
DCK
Full length human recombit protein of human DCK produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Western Blot
OTI16E12
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P27707
deoxycytidine kinase
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1633