Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCK Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: 10412, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243639V

Catalog No. NBP243639V


Add to cart

Description

Description

DCK Monoclonal antibody specifically detects DCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

DCK
10412
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
DCK
Recombinant protein encompassing a sequence within the center region of human DCK. The exact sequence is proprietary.
Protein A purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
dCK, deoxycytidine kinase, EC 2.7.1, EC 2.7.1.74, MGC117410, MGC138632
Mouse
IgG2b
0.1 ml
Primary
1633
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit