Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCK Mouse anti-Human, Clone: OTI16G6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525502

Catalog No. PIMA525502


Add to cart

Description

Description

Deoxycytidine kinase is responsible for the phosphorylation of several deoxyribonucleosides and their analogs. Deficiency of this enzyme activity is associated with resistance to antiviral and anticancer chemotherapeutic agents, whereas increased enzyme activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. It is the rate limiting enzyme in the activation of many important anticancer and retroviral drugs and its activity is often decreased in cells that are resistant to cytosine arabinoside.
Specifications

Specifications

DCK
Monoclonal
0.49 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P27707
Deoxycytidine kinase; dCK
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1633
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI16G6
Unconjugated
DCK
Liquid
DCK
Full length human recombit protein of DCK produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit