Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCLRE1B Mouse anti-Human, Clone: OTI2B7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502352

Catalog No. 50-167-3414


Add to cart

Description

Description

DNA interstrand cross-links prevent strand separation, thereby physically blocking transcription, replication, and segregation of DNA. DCLRE1B is one of several evolutionarily conserved genes involved in repair of interstrand cross-links.
Specifications

Specifications

DCLRE1B
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H816
APOLLO, SNM1B, SNMIB
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64858
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2B7
Unconjugated
DCLRE1B
Liquid
DCLRE1B
Human recombit protein fragment corresponding to amino acids 1-280 of human DCLRE1B produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit