Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCLRE1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP257379
Description
DCLRE1B Polyclonal specifically detects DCLRE1B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DCLRE1B | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
APOLLO, DKFZp686C24187, DNA cross-link repair 1B, DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae), DNA cross-link repair 1B protein, EC 3.1, FLJ12810, FLJ13998, hSNM1B, PSO2 homolog, SNM1 homolog B, SNM1B5' exonuclease Apollo, SNMIB | |
Rabbit | |
Affinity Purified | |
RUO | |
64858 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
DCLRE1B | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VVPIVSRRPCGGFQDSLSPRISVPLIPDSVQQYMSSSSRKPSLLWLLERRLKRPRTQGVVFESPEESADQSQADRDSKKAKKEKLSPWPADLEKQPSHHPLRIKKQ | |
100 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction