Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCLRE1B Mouse anti-Human, Clone: OTI2B7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525400

Catalog No. PIMA525400


Add to cart

Description

Description

DNA interstrand cross-links prevent strand separation, thereby physically blocking transcription, replication, and segregation of DNA. DCLRE1B is one of several evolutionarily conserved genes involved in repair of interstrand cross-links.
Specifications

Specifications

DCLRE1B
Monoclonal
0.62 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H816
APOLLO, SNM1B, SNMIB, 5′ exonuclease Apollo, DNA cross-link repair 1B (PSO2 homolog, S. cerevisiae), PSO2 homolog, SNM1 homolog B
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
64858
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2B7
Unconjugated
DCLRE1B
Liquid
DCLRE1B
Human recombit protein fragment corresponding to amino acids 1-280 of DCLRE1B produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit