Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DCP1B Mouse anti-Human, Clone: OTI1B5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527231

Catalog No. PIMA527231


Add to cart

Description

Description

DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.
Specifications

Specifications

DCP1B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IZD4
DCP1, hDcp1b, decapping enzyme hDcp1b, mRNA-decapping enzyme 1B
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
196513
Western Blot
OTI1B5
Unconjugated
DCP1B
Liquid
DCP1B
Human recombit protein fragment corresponding to amino acids 167-473 of DCP1B produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit