Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCP1B Monoclonal Antibody (OTI1C5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527230
Description
DCP1B Monoclonal antibody specifically detects DCP1B in Human samples. It is validated for Western Blot.Specifications
DCP1B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IZD4 | |
DCP1B | |
Human recombinant protein fragment corresponding to amino acids 167-473 of DCP1B produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI1C5 | |
Unconjugated | |
DCP1B | |
B930050E02Rik; DCP1; DCP1 decapping enzyme homolog b; DCP1 decapping enzyme homolog B (S. cerevisiae); Dcp1b; decapping enzyme hDcp1b; decapping mRNA 1B; hDcp1b; mRNA-decapping enzyme 1B; RGD1562214 | |
Mouse | |
Affinity Chromatography | |
RUO | |
196513 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction