Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DCP2 Antibody (PCRP-DCP2-1D6), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314037AF488
This item is not returnable.
View return policy
Description
DCP2 Monoclonal antibody specifically detects DCP2 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
DCP2 | |
Monoclonal | |
Alexa Fluor 488 | |
DCP2 decapping enzyme homolog (S. cerevisiae), EC 3.-, FLJ33245, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20, Nudix motif 20, NUDT20Nucleoside diphosphate-linked moiety X motif 20 | |
Recombinant full-length human DCP2protein (Uniprot: Q8IU60) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
PCRP-DCP2-1D6 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
167227 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction