Learn More
Invitrogen™ DCTD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534947
Description
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH, human DCTD-transfected 293T cells. Predicted reactivity: Mouse (94%), Rat (94%), Zebrafish (90%), Xenopus laevis (90%), Chicken (90%), Rhesus Monkey (97%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene catalyzes the deamination of dCMP to dUMP, the nucleotide substrate for thymidylate synthase. The encoded protein is allosterically activated by dCTP and inhibited by dTTP, and is found as a homohexamer. This protein uses zinc as a cofactor for its activity. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
DCTD | |
Polyclonal | |
Unconjugated | |
DCTD | |
6030466N05Rik; dCMP deaminase; Dctd; Deoxycytidylate deaminase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P32321 | |
DCTD | |
Recombinant fragment contains a sequence corresponding to a region within amino acids 1 and 178 of DCTD. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.