Learn More
Aviva Systems Biology Corp DCUN1D3 Antibody - N-terminal region (ARP53230_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53230_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DCUN1D3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DCUN1D3. Alternate names: 44M2.4, DKFZp686O0290, FLJ41725, MGC48972ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-60980